Learn More
Microsolv Technology Corporation Caps, Screw Top, with White Silicone Rubber / Natural PTFE Septa, fitted in "knurled", polypropylene, Purple Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.
Supplier:  Microsolv Technology Corporation 9502SE15PM
Encompass_Preferred
This is our most economical, lowest cost Screw Cap for Autosampler Vials.Natural PTFE is as equally inert and durable as Red PTFE, but no dyes have been added.
Catalog No. 50-227-0482