Learn More
Microsolv Technology Corporation Caps, Screw Top, with Natural Silicone Rubber / White PTFE Septa, bonded in "knurled", polypropylene, Blue Caps. For use with 9-425 thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271286
Encompass_Preferred
Natural Silicone Rubber is made without adding color and has the same properties as White Silicone Rubber and is a less expensive bonded Septa and Cap.
This item is not returnable. View return policy
Catalog No. 50-227-1286 Supplier Microsolv Technology Corporation Supplier No. 9502S11CB
Natural Silicone Rubber is made without adding color and has the same properties as White Silicone Rubber and is a less expensive bonded Septa and Cap.