Learn More
Microsolv Technology Corporation Caps, Screw Top, with White Silicone Rubber / Natural TEF Septa, fitted in smooth sided, Robo Style, soft polypropylene, Blue Caps. For use with 9-425 thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270296
Encompass_Preferred
White silicone rubber and white TEF fitted septa for a lower cost cap. TEF is equally inert as PTFE for standard chromatography.
This item is not returnable. View return policy
Catalog No. 50-227-0296 Supplier Microsolv Technology Corporation Supplier No. 9502S10TEF
White silicone rubber and white TEF fitted septa for a lower cost cap. TEF is equally inert as PTFE for standard chromatography.