Learn More
Microsolv Technology Corporation Caps, Screw Top, with White Silicone Rubber / Red PTFE Septa / Liner, fitted in knurled polypropylene, Black Caps. For use with ND15 thread, storage Vials or Bottles. 1000/CS.
Supplier:  Microsolv Technology Corporation 9701510KK
Encompass_Preferred
Silicone Rubber / PTFE Septa is 45⁰ Shore A (hardness) and is 1.3mm thick. Use as septa or a liner. Cap has a 9mm center hole. Can be used for use in Non-Critical Analytical Testing and is intended for storage. Use with Vials or Bottles with 15-400mm screw threads.
Catalog No. 50-227-1331