Learn More
Microsolv Technology Corporation Caps, Screw Top, with White Silicone Rubber / White PTFE, ultra-low bleed Septa, bonded in "knurled", polypropylene, Dark Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK
Catalog No. 502270485
Encompass_Preferred
This very high purity Septa has low extractables and is excellent choice for HPLC. Compares to "Certified" Brands.
This item is not returnable. View return policy
Catalog No. 50-227-0485 Supplier Microsolv Technology Corporation Supplier No. 9509S10B
This very high purity Septa has low extractables and is excellent choice for HPLC. Compares to "Certified" Brands.