Learn More
Microsolv Technology Corporation Caps, Screw Top, with White Silicone Rubber / White PTFE Septa, fitted in "knurled", polypropylene, Green Caps. Recommended for Mass Spec. For use with 9-425mm thread, 12x32mm autosampler vials. Compares to Agilent caps. 100/PK.
Catalog No. 502270298
Encompass_Preferred
Made with ultra-pure silicone/PTFE septa and recommended for LCMS or GCMS.
This item is not returnable. View return policy
Catalog No. 50-227-0298 Supplier Microsolv Technology Corporation Supplier No. 9502S10MG
Made with ultra-pure silicone/PTFE septa and recommended for LCMS or GCMS.