Learn More
Microsolv Technology Corporation Caps, Screw Top, with White Silicone Rubber / Natural PTFE Septa, fitted in "knurled", polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502270480
Encompass_Preferred
This is our most economical, lowest cost Screw Cap for Autosampler Vials. Natural PTFE is as equally inert and durable as Red PTFE, but no dyes have been added.
This item is not returnable. View return policy
Catalog No. 50-227-0480 Supplier Microsolv Technology Corporation Supplier No. 9502SE15KM
This is our most economical, lowest cost Screw Cap for Autosampler Vials. Natural PTFE is as equally inert and durable as Red PTFE, but no dyes have been added.