Learn More
Microsolv Technology Corporation Caps, Screw Top, with Silicone Rubber / PTFE Partial Pre-Slit (Y Shape) Septa, fitted in "knurled" polypropylene, Blue Caps. For use with 9-425 thread, 12x32mm autosampler vials. Excellent for GC and GCMS. 1000/CS.
Catalog No. 502271303
Encompass_Preferred
Partial slit, only in the rubber leaves PTFE to form an evaporation barrier making this a good choice for volatile samples when an autosampler requires a pre-slit Septa.Y shaped partial slit allows for better "resealing" of rubber after initial injection.
This item is not returnable. View return policy
Catalog No. 50-227-1303 Supplier Microsolv Technology Corporation Supplier No. 9502S34CBM
Partial slit, only in the rubber leaves PTFE to form an evaporation barrier making this a good choice for volatile samples when an autosampler requires a pre-slit Septa.Y shaped partial slit allows for better "resealing" of rubber after initial injection.