Learn More
Microsolv Technology Corporation Caps, Screw Top, with White Silicone Rubber / White PTFE, ultra-low bleed, Pre-Slit Septa, bonded in "knurled", polypropylene, Dark Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK
Catalog No. 502270166
Encompass_Preferred
Pre-slit for easy penetration by autosampler needles and recommended for Waters Alliance, Acquity™ and all other autosamplers that require Pre-Slit Septa. Good choice for LCMS & LCMS/MS.
This item is not returnable. View return policy
Catalog No. 50-227-0166 Supplier Microsolv Technology Corporation Supplier No. 9509S30B
Pre-slit for easy penetration by autosampler needles and recommended for Waters Alliance, Acquity™ and all other autosamplers that require Pre-Slit Septa. Good choice for LCMS & LCMS/MS.