Learn More
Microsolv Technology Corporation Caps Screw Top 9Mm Closedcaps
Catalog No. 5001452090
Encompass_Preferred
Caps, Screw Top, Solid, with a 0.2mm thick, PTFE liner, in knurled, closed Blue Caps. For use with 9-425mm thread,12x32mm autosampler vials. MicroSolv Brand 100/PK.
Catalog No. 50-014-52090 Supplier Microsolv Technology Corporation Supplier No. 9502S09ST
Caps, Screw Top, Solid, with a 0.2mm thick, PTFE liner, in knurled, closed Blue Caps. For use with 9-425mm thread,12x32mm autosampler vials. MicroSolv Brand 100/PK.