Learn More
Microsolv Technology Corporation Caps Screw Top Green Bondedca
Catalog No. 502766
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in knurled, polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 1000/CS.
Catalog No. 50-276-6 Supplier Microsolv Technology Corporation Supplier No. 9502S10CGM
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in knurled, polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 1000/CS.