Learn More
Microsolv Technology Corporation Caps, Screw Top, with Orange Red Rubber / PTFE Septa, bonded in "knurled", polypropylene, Red Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271291
Encompass_Preferred
Orange-Red Silicone Rubber is more economical White Silicone Rubber and softer than Natural Red Rubber which is better for your autosamplers, minimizing damaging needles and injections systems.
This item is not returnable. View return policy
Catalog No. 50-227-1291 Supplier Microsolv Technology Corporation Supplier No. 9502S40CR
Orange-Red Silicone Rubber is more economical White Silicone Rubber and softer than Natural Red Rubber which is better for your autosamplers, minimizing damaging needles and injections systems.