Learn More
Microsolv Technology Corporation Caps, Screw Top, with Orange Silicone Rubber / Ivory PTFE, low durometer Septa, fitted in smooth sided, Robo Style, soft polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271293
Encompass_Preferred
Orange-Red Silicone Rubber is softer than Natural Red Rubber and is better for your autosamplers. Ivory white is inert. Compares to National Scientific / Thermo DP™ Screw Caps.
This item is not returnable. View return policy
Catalog No. 50-227-1293 Supplier Microsolv Technology Corporation Supplier No. 9502S01119
Orange-Red Silicone Rubber is softer than Natural Red Rubber and is better for your autosamplers. Ivory white is inert. Compares to National Scientific / Thermo DP™ Screw Caps.