Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, without a liner. "knurled", polypropylene, Black Caps. For use with 13-425mm thread, 15x45mm autosampler vials. 100/PK.
Catalog No. 502269942
Encompass_Preferred
This cap is not recommended for storage or transportation of chemicals or samples to be analyzed. Good for storage of dry objects or after inserting a liner.
This item is not returnable. View return policy
Catalog No. 50-226-9942 Supplier Microsolv Technology Corporation Supplier No. 9504S01CBK
This cap is not recommended for storage or transportation of chemicals or samples to be analyzed. Good for storage of dry objects or after inserting a liner.