Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with a Red-Orange Silicone Rubber / TEF liner, in "knurled", closed Blue Caps. For use with 9-425mm thread,12x32mm autosampler vials. 100/PK.
Catalog No. 502271271
Encompass_Preferred
Silicone Rubber with TEF liner is inert lowers the cost. Liner is 53° Shore D, 0-2mm. Solid top turns autosampler vials into storage vials.
This item is not returnable. View return policy
Catalog No. 50-227-1271 Supplier Microsolv Technology Corporation Supplier No. 9502S09SR
Silicone Rubber with TEF liner is inert lowers the cost. Liner is 53° Shore D, 0-2mm. Solid top turns autosampler vials into storage vials.