Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with Natural Silicone Rubber / Natural PTFE Liner, fitted in knurled polypropylene, White Caps. For use with 24-414mm threaded EPA Style vials. ND24 threads. 100/PK.
Catalog No. 502270978
Encompass_Preferred
Liner is 3.18mm (0.125") thick and can be used for long term storage with any 30, 40 or 60 EPA Style vial that has 40-414 threads. Solid top cap does not have an opening and is designed for storage or transportation.
This item is not returnable. View return policy
Catalog No. 50-227-0978 Supplier Microsolv Technology Corporation Supplier No. 97041SP24
Liner is 3.18mm (0.125") thick and can be used for long term storage with any 30, 40 or 60 EPA Style vial that has 40-414 threads. Solid top cap does not have an opening and is designed for storage or transportation.