Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with a Red Orange Natural Rubber/ Transparent TFE liner, in "knurled", polypropylene, Black Caps. For use with 13-425mm thread,15x45mm autosampler vials. 100/PK.
Catalog No. 502269939
Encompass_Preferred
This Solid Cap turns 15x45mm, 4ml autosampler vials into storage bottles. Rubber and TFE liner provide an inert seal.
This item is not returnable. View return policy
Catalog No. 50-226-9939 Supplier Microsolv Technology Corporation Supplier No. 9504S80CBK
This Solid Cap turns 15x45mm, 4ml autosampler vials into storage bottles. Rubber and TFE liner provide an inert seal.