Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with a Silicone Rubber / Polypropylene liner, in "knurled", closed Black Caps. 1000/CS
Catalog No. 502269941
Encompass_Preferred
This item has a minimum order quantity of 20 packs per supplier requirements.
Polyfluoroalkyl Substances (PFAS) can be tested or stored in our autosampler vials using these caps. The silicone rubber (for optimal sealing), and polypropylene fitted liners (for inertness) are excellent for storage of your sample before or after chromatography. These caps do not contain PTFE.
This item is not returnable. View return policy
Catalog No. 50-226-9941 Supplier Microsolv Technology Corporation Supplier No. 9502S09PSM
This item has a minimum order quantity of 20 packs per supplier requirements.
Polyfluoroalkyl Substances (PFAS) can be tested or stored in our autosampler vials using these caps. The silicone rubber (for optimal sealing), and polypropylene fitted liners (for inertness) are excellent for storage of your sample before or after chromatography. These caps do not contain PTFE.