Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with Silicone Rubber / PTFE Liner, fitted in knurled polypropylene, White Caps. For use with ND20 thread, storage Vials or Bottles. 100/PK.
Catalog No. 502271275
Encompass_Preferred
Silicone Rubber / PTFE Liner can be used for use with Storage Vials / Bottles with 20-400mm screw threads.
This item is not returnable. View return policy
Catalog No. 50-227-1275 Supplier Microsolv Technology Corporation Supplier No. 98100SP20W
Silicone Rubber / PTFE Liner can be used for use with Storage Vials / Bottles with 20-400mm screw threads.