Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with a Silicone Rubber / PTFE liner, in "knurled", polypropylene, Black Caps. For use with 13-425mm thread,15x45mm autosampler vials. 100/PK.
Catalog No. 502270426
Encompass_Preferred
Turns 4ml, 15x45 autosampler vials into storage bottles. The Silicone Rubber / PTFE liner provides an inert surface with good sealing properties.
This item is not returnable. View return policy
Catalog No. 50-227-0426 Supplier Microsolv Technology Corporation Supplier No. 98100SP13K
Turns 4ml, 15x45 autosampler vials into storage bottles. The Silicone Rubber / PTFE liner provides an inert surface with good sealing properties.