Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with Silicone Rubber / PTFE Liner, fitted in knurled polypropylene, Black Caps. For use with ND15 thread, storage Vials or Bottles. 100/PK.
Catalog No. 502270428
Encompass_Preferred
Silicone Rubber / PTFE Liner can be used for use with Storage Vials or Bottles with 15-415mm screw threads. Also, used with heavy wall reaction vials.
This item is not returnable. View return policy
Catalog No. 50-227-0428 Supplier Microsolv Technology Corporation Supplier No. 98100SP15K
Silicone Rubber / PTFE Liner can be used for use with Storage Vials or Bottles with 15-415mm screw threads. Also, used with heavy wall reaction vials.