Learn More
Microsolv Technology Corporation Caps, Screw Top, with an Open Hole. "knurled", polypropylene, Black Caps. Septa are sold separately and must be inserted manually. For use with 13-425mm thread, 15x45mm autosampler vials. 1000/CS.
Catalog No. 502271280
Encompass_Preferred
Caps, Screw Top, with an Open Hole. "knurled", polypropylene, Black Caps. Septa are sold separately and must be inserted manually. For use with 13-425mm thread, 15x45mm autosampler vials. 1000/CS.
This item is not returnable. View return policy
Catalog No. 50-227-1280 Supplier Microsolv Technology Corporation Supplier No. 9504S00CBKM
Caps, Screw Top, with an Open Hole. "knurled", polypropylene, Black Caps. Septa are sold separately and must be inserted manually. For use with 13-425mm thread, 15x45mm autosampler vials. 1000/CS.