Learn More
Microsolv Technology Corporation Caps, Screw Top, with an Open Hole. "Knurled", polypropylene, Black Caps. Septa are sold separately and must be inserted manually. For use with 8-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270431
Encompass_Preferred
Caps, Screw Top, with an Open Hole. "Knurled", polypropylene, Black Caps. Septa are sold separately and must be inserted manually. For use with 8-425mm thread, 12x32mm autosampler vials. 100/PK.
This item is not returnable. View return policy
Catalog No. 50-227-0431 Supplier Microsolv Technology Corporation Supplier No. 9582SCAP1
Caps, Screw Top, with an Open Hole. "Knurled", polypropylene, Black Caps. Septa are sold separately and must be inserted manually. For use with 8-425mm thread, 12x32mm autosampler vials. 100/PK.