Learn More
Microsolv Technology Corporation Caps, Screw Top, with an Open Hole. "Knurled", polypropylene, Blue Caps. Septa are sold separately and must be inserted manually. For use with 9-425mm thread, 12x32mm autosampler vials. 1,000/CS.
Catalog No. 502270432
Encompass_Preferred
These caps are used for our Soft-Guard Brand Caps.
This item is not returnable. View return policy
Catalog No. 50-227-0432 Supplier Microsolv Technology Corporation Supplier No. 9502S01BM
These caps are used for our Soft-Guard Brand Caps.