Learn More
Microsolv Technology Corporation Caps,Screwcap,W Slit/Ptfe C/Pk
Catalog No. NC1344222
Encompass_Preferred
Caps, Screw Top, with Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. Soft-Guard Brand. 100/PK
Catalog No. NC1344222 Supplier Microsolv Technology Corporation Supplier No. 9502SE30CB
Caps, Screw Top, with Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. Soft-Guard Brand. 100/PK