Learn More
Microsolv Technology Corporation Caps Scrw Aqr Light Blue Wit
Catalog No. 5001452288
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE, ultra-pure Septa, fitted in knurled, polypropylene, Light Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. AQR Brand. 100/PK.
Catalog No. 50-014-52288 Supplier Microsolv Technology Corporation Supplier No. 9509S10CB
Caps, Screw Top, with Silicone Rubber / PTFE, ultra-pure Septa, fitted in knurled, polypropylene, Light Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. AQR Brand. 100/PK.