Learn More
Microsolv Technology Corporation Caps Scrw Red Preslit Sil/Ptfe
Catalog No. 502773
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Red Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 1000/CS
Catalog No. 50-277-3 Supplier Microsolv Technology Corporation Supplier No. 9502S30CRM
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Red Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 1000/CS