Learn More
Microsolv Technology Corporation Caps Scrw Sil/Ptfe Bluecaps
Catalog No. 5001452108
Encompass_Preferred
Caps, Screw Top, with ultra-clean Silicone Rubber / PTFE Septa, fitted in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK
Catalog No. 50-014-52108 Supplier Microsolv Technology Corporation Supplier No. 9502S14CB
Caps, Screw Top, with ultra-clean Silicone Rubber / PTFE Septa, fitted in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK