Learn More
Microsolv Technology Corporation Caps, Snap Top, with 0.04" thick, White Silicone Rubber / Natural PTFE Septa, fitted in smooth, polyethylene, Yellow Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Catalog No. 502270396
Encompass_Preferred
Natural PTFE is very pure and does not have dyes that could extract from it.Our most economical Silicone Rubber / PTFE Snap Cap.
This item is not returnable. View return policy
Catalog No. 50-227-0396 Supplier Microsolv Technology Corporation Supplier No. 9502CE15YM
Natural PTFE is very pure and does not have dyes that could extract from it.Our most economical Silicone Rubber / PTFE Snap Cap.