Learn More
Microsolv Technology Corporation Caps, Snap Top, with 0.04" thick, White Silicone Rubber / Yellow PTFE Pre-Slit (Star Shape) Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502270423
Encompass_Preferred
Ultra-low Bleed Septa provide less extractables than most septa and is recommended for LCMS. The star shaped (*) slit offers a much greater target area for an autosampler needle to penetrate the septa and offers minimal resistance. It may not reseal as well as other Pre-Slit Septa.
This item is not returnable. View return policy
Catalog No. 50-227-0423 Supplier Microsolv Technology Corporation Supplier No. 9502CE31CB
Ultra-low Bleed Septa provide less extractables than most septa and is recommended for LCMS. The star shaped (*) slit offers a much greater target area for an autosampler needle to penetrate the septa and offers minimal resistance. It may not reseal as well as other Pre-Slit Septa.