Learn More
Microsolv Technology Corporation Caps, Snap Top, with 0.04" thick, White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Catalog No. 502270488
Encompass_Preferred
Pre-Slit Septa are normally recommended for Waters Alliance and Acquity instruments. Natural Septa does not have any color pigment and will not introduce extractables.
This item is not returnable. View return policy
Catalog No. 50-227-0488 Supplier Microsolv Technology Corporation Supplier No. 9502CE35GM
Pre-Slit Septa are normally recommended for Waters Alliance and Acquity instruments. Natural Septa does not have any color pigment and will not introduce extractables.