Learn More
Microsolv Technology Corporation Caps, Snap Top, with 0.04" thick, White Silicone Rubber / Natural PTFE Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502270391
Encompass_Preferred
Natural PTFE is very pure and does not have dyes that could extract from it. Our most economical Silicone Rubber / PTFE Snap Cap.
This item is not returnable. View return policy
Catalog No. 50-227-0391 Supplier Microsolv Technology Corporation Supplier No. 9502CE15G
Natural PTFE is very pure and does not have dyes that could extract from it. Our most economical Silicone Rubber / PTFE Snap Cap.