Learn More
Microsolv Technology Corporation Caps, Snap Top, with 1.3mm thick, Silicone Rubber / PTFE Septa, fitted in smooth, soft polyethylene, Natural Translucent Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502270301
Encompass_Preferred
Soft polyethylene caps are more pliable than other Snap Caps and are easier to place on the vial. Popular choice when using a large number of caps at one time. These caps may not seal as well as other Snap Caps and should be used with sample solvents with low volatility.
This item is not returnable. View return policy
Catalog No. 50-227-0301 Supplier Microsolv Technology Corporation Supplier No. 9502CS10T
Soft polyethylene caps are more pliable than other Snap Caps and are easier to place on the vial. Popular choice when using a large number of caps at one time. These caps may not seal as well as other Snap Caps and should be used with sample solvents with low volatility.