Learn More
Microsolv Technology Corporation Caps, Snap Top, with 1.00mm thick, Red PTFE / White Silicone Rubber / Red PTFE Septa, fitted in smooth, soft polyethylene, Natural Translucent Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502271332
Encompass_Preferred
Soft polyethylene caps are more pliable than other Snap Caps and are easier to place on the vial. Popular choice when using a large number of caps at one time. These caps may not seal as well as other Snap Caps and should be used with sample solvents with low volatility. This septa was designed for specific, older instruments such as Agilent 1050 and to provide extra cleaning when "wiping" a needle. Also provides a small degree of better resealing after injection compared to Silicone Rubber / PTFE Septa.
This item is not returnable. View return policy
Catalog No. 50-227-1332 Supplier Microsolv Technology Corporation Supplier No. 9502CS20T
Soft polyethylene caps are more pliable than other Snap Caps and are easier to place on the vial. Popular choice when using a large number of caps at one time. These caps may not seal as well as other Snap Caps and should be used with sample solvents with low volatility. This septa was designed for specific, older instruments such as Agilent 1050 and to provide extra cleaning when "wiping" a needle. Also provides a small degree of better resealing after injection compared to Silicone Rubber / PTFE Septa.