Learn More
Microsolv Technology Corporation Caps, Snap Top, with 1.0mm thick, Orange Rubber / Transparent TEF Septa, fitted in Smooth, soft polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502271337
Encompass_Preferred
Caps, Snap Top, with 1.0mm thick, Orange Rubber / Transparent TEF Septa, fitted in Smooth, soft polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
This item is not returnable. View return policy
Catalog No. 50-227-1337 Supplier Microsolv Technology Corporation Supplier No. 9502CS60B
Caps, Snap Top, with 1.0mm thick, Orange Rubber / Transparent TEF Septa, fitted in Smooth, soft polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.