Learn More
Microsolv Technology Corporation Caps, Snap Top, with 1.00mm thick, White Silicone Rubber / Red PTFE Pre-Slit (Y Shape) Septa, fitted in smooth, soft polyethylene, Natural Transparent Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502270299
Encompass_Preferred
Soft polyethylene caps are more pliable than other Snap Caps and are easier to place on the vial. Popular choice when using a large number of caps at one time. These caps may not seal as well as other Snap Caps and should be used with sample solvents with low volatility. Y Shape Pre-Slit provides a greater target area for needles and are easier to penetrate. The 1.0mm Septa is thicker than most and offers improved sealing compared with other similar Septa. Pre-Slit Septa are normally recommended for Waters Alliance and Acquity instruments.
This item is not returnable. View return policy
Catalog No. 50-227-0299 Supplier Microsolv Technology Corporation Supplier No. 9502CS34T
Soft polyethylene caps are more pliable than other Snap Caps and are easier to place on the vial. Popular choice when using a large number of caps at one time. These caps may not seal as well as other Snap Caps and should be used with sample solvents with low volatility. Y Shape Pre-Slit provides a greater target area for needles and are easier to penetrate. The 1.0mm Septa is thicker than most and offers improved sealing compared with other similar Septa. Pre-Slit Septa are normally recommended for Waters Alliance and Acquity instruments.