Learn More
Microsolv Technology Corporation Caps Snap Blue Sil/Ptfe Septa
Catalog No. 5001451999
Encompass_Preferred
Caps, Snap Top, with Silicone Rubber / PTFE Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. BASIK Brand. 1000/CS.
Catalog No. 50-014-51999 Supplier Microsolv Technology Corporation Supplier No. 9502CE05BM
Caps, Snap Top, with Silicone Rubber / PTFE Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. BASIK Brand. 1000/CS.