Learn More
Microsolv Technology Corporation Caps Snap Blue Sil/Ptfecaps
Catalog No. 5001451923
Encompass_Preferred
Caps, Snap Top, with Silicone Rubber / PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 1000/CS.
Catalog No. 50-014-51923 Supplier Microsolv Technology Corporation Supplier No. 9502C10CBM
Caps, Snap Top, with Silicone Rubber / PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 1000/CS.