Learn More
Microsolv Technology Corporation Caps, Snap Top, with Butyl Rubber / Tefzel Septa, fitted in smooth, soft polyethylene, Natural Transparent Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502271341
Encompass_Preferred
Soft polyethylene caps are more pliable than other Snap Caps and are easier to place on the vial. Popular choice when using a large number of caps at one time. These caps may not seal as well as other Snap Caps and should be used with sample solvents with low volatility. Butyl Rubber with Tefzel is a lower cost Septa but inert enough for most applications.
This item is not returnable. View return policy
Catalog No. 50-227-1341 Supplier Microsolv Technology Corporation Supplier No. 9502CS08T
Soft polyethylene caps are more pliable than other Snap Caps and are easier to place on the vial. Popular choice when using a large number of caps at one time. These caps may not seal as well as other Snap Caps and should be used with sample solvents with low volatility. Butyl Rubber with Tefzel is a lower cost Septa but inert enough for most applications.