Learn More
Microsolv Technology Corporation Caps Snap Gr Preslit Sil/Ptfe
Catalog No. 5001451952
Encompass_Preferred
Caps, Snap Top, with Silicone Rubber / PTFE Pre-Slit Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 1000/CS.
Catalog No. 50-014-51952 Supplier Microsolv Technology Corporation Supplier No. 9502C30CGM
Caps, Snap Top, with Silicone Rubber / PTFE Pre-Slit Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 1000/CS.