Learn More
Microsolv Technology Corporation Caps, Snap Top, with Silicone Rubber / PTFE Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502270374
Encompass_Preferred
Our most economical Silicone Rubber / PTFE Snap Cap.
This item is not returnable. View return policy
Catalog No. 50-227-0374 Supplier Microsolv Technology Corporation Supplier No. 9502CE05B
Our most economical Silicone Rubber / PTFE Snap Cap.