Learn More
Microsolv Technology Corporation Caps Snap Soft Preslit X
Catalog No. 502759
Encompass_Preferred
Caps, Snap Top, with 1.00mm thick, White Silicone Rubber / Blue PTFE Pre-Slit (X Shape) Septa, fitted in smooth, soft polyethylene, Natural Transparent Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 1000/CS.
Catalog No. 50-275-9 Supplier Microsolv Technology Corporation Supplier No. 9502CS30TM
Caps, Snap Top, with 1.00mm thick, White Silicone Rubber / Blue PTFE Pre-Slit (X Shape) Septa, fitted in smooth, soft polyethylene, Natural Transparent Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 1000/CS.