Learn More
Microsolv Technology Corporation Caps Snap Top Blue White Ptfe
Catalog No. 5001451984
Encompass_Preferred
Caps, Snap Top, with White PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51984 Supplier Microsolv Technology Corporation Supplier No. 9502C50CB
Caps, Snap Top, with White PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 100/PK.