Learn More
Microsolv Technology Corporation Caps Snap Top Natural Sil/Ptfe
Catalog No. 5001451928
Encompass_Preferred
Caps, Snap Top, with Silicone Rubber / PTFE Septa, fitted in smooth, polyethylene, Natural Translucent Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51928 Supplier Microsolv Technology Corporation Supplier No. 9502C10CN
Caps, Snap Top, with Silicone Rubber / PTFE Septa, fitted in smooth, polyethylene, Natural Translucent Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. MicroSolv Brand. 100/PK.