Learn More
Microsolv Technology Corporation Caps, Snap Top, with Red PTFE / White Silicone Rubber / Red PTFE Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502271233
Encompass_Preferred
This septa was designed for specific, older instruments such as Agilent 1050 and to provide extra cleaning when "wiping" a needle. Also provides a small degree of better resealing after injection compared to Silicone Rubber / PTFE Septa.
This item is not returnable. View return policy
Catalog No. 50-227-1233 Supplier Microsolv Technology Corporation Supplier No. 9502C20CG
This septa was designed for specific, older instruments such as Agilent 1050 and to provide extra cleaning when "wiping" a needle. Also provides a small degree of better resealing after injection compared to Silicone Rubber / PTFE Septa.