Learn More
Microsolv Technology Corporation Caps, Snap Top, with Red PTFE / White Silicone Rubber / Red PTFE Septa, fitted in smooth, polyethylene, Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Catalog No. 502271232
Encompass_Preferred
This septa was designed for specific, older instruments such as Agilent 1050 and to provide extra cleaning when "wiping" a needle. Also provides a small degree of better resealing after injection compared to Silicone Rubber / PTFE Septa. Buying in case quantities can save money and time.
This item is not returnable. View return policy
Catalog No. 50-227-1232 Supplier Microsolv Technology Corporation Supplier No. 9502C20CBM
This septa was designed for specific, older instruments such as Agilent 1050 and to provide extra cleaning when "wiping" a needle. Also provides a small degree of better resealing after injection compared to Silicone Rubber / PTFE Septa. Buying in case quantities can save money and time.