Learn More
Microsolv Technology Corporation Caps, Snap Top, with Red Rubber / PTFE Septa, fitted in smooth, polyethylene, Yellow Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502271251
Encompass_Preferred
Red Rubber / PTFE is often the choice of labs that do not want to analyze with silicone rubber in the Septa and want a lower cost cap.
This item is not returnable. View return policy
Catalog No. 50-227-1251 Supplier Microsolv Technology Corporation Supplier No. 9502C40CY
Red Rubber / PTFE is often the choice of labs that do not want to analyze with silicone rubber in the Septa and want a lower cost cap.