Learn More
Microsolv Technology Corporation Caps, Snap Top, with Silicone Rubber / PTFE Pre-Slit Septa, fitted in smooth, polyethylene, Yellow Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 1000/CS.
Catalog No. 502269923
Encompass_Preferred
This is one of the snap caps on the market and has been available for over 25 years. Pre-Slit septa are normally recommended for Waters Alliance instruments.
This item is not returnable. View return policy
Catalog No. 50-226-9923 Supplier Microsolv Technology Corporation Supplier No. 9502C30CYM
This is one of the snap caps on the market and has been available for over 25 years. Pre-Slit septa are normally recommended for Waters Alliance instruments.