Learn More
Microsolv Technology Corporation Caps, Snap Top, with Silicone Rubber / PTFE Pre-Slit Septa, fitted in smooth, polyethylene, Green Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502270363
Encompass_Preferred
This is one of the snap caps on the market and has been available for over 25 years. Pre-Slit septa are normally recommended for Waters Alliance instruments.
This item is not returnable. View return policy
Catalog No. 50-227-0363 Supplier Microsolv Technology Corporation Supplier No. 9502C30CG
This is one of the snap caps on the market and has been available for over 25 years. Pre-Slit septa are normally recommended for Waters Alliance instruments.