Learn More
Microsolv Technology Corporation Caps, Snap Top, with Silicone Rubber / PTFE Pre-Slit (X Shape) Septa, fitted in smooth, polyethylene, Yellow Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Catalog No. 502271241
Encompass_Preferred
This is one of the snap caps on the market and has been available for over 25 years. Pre-Slit septa are normally recommended for Waters Alliance instruments. X Shape Pre-Slit provides a greater target area for needles and are easier to penetrate but do not reseal as well as single slit septa.
This item is not returnable. View return policy
Catalog No. 50-227-1241 Supplier Microsolv Technology Corporation Supplier No. 9502C30CYX
This is one of the snap caps on the market and has been available for over 25 years. Pre-Slit septa are normally recommended for Waters Alliance instruments. X Shape Pre-Slit provides a greater target area for needles and are easier to penetrate but do not reseal as well as single slit septa.